Läs mer
N-[3-[3-(Dimethylamino)-1-oxo-2-propenyl]phenyl]-N-ethylacetamide, Mikromol™
Produktkod. 30462393
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462393 4 x 25 mg
30051565 25 mg
30051928 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30462393 Leverantör Mikromol Leverantörsnummer MM1322.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[3-[3-(Dimethylamino)-1-oxo-2-propenyl]phenyl]-N-ethylacetamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[3-[3-(Dimethylamino)-1-oxo-2-propenyl]phenyl]-N-ethylacetamide
CAS 96605-66-2
Analyt- eller komponentnamn N-[3-[3-(Dimethylamino)-1-oxo-2-propenyl]phenyl]-N-ethylacetamide
Frakt skick Room Temperature
Molekylformel C15 H20 N2 O2
Certifikat och efterlevnad ISO 17025
Synonym N-[3-[3-(Dimethylamino)-1-oxo-2-propenyl]phenyl]-N-ethylacetamide, Zaleplon Related Compound A
InChI formel InChI=1S/C15H20N2O2/c1-5-17(12(2)18)14-8-6-7-13(11-14)15(19)9-10-16(3)4/h6-11H,5H2,1-4H3/b10-9+
Lösningstyp Neat
LEDER CCN(C(=O)C)c1cccc(c1)C(=O)\C=C\N(C)C
IUPAC-namn N-[3-[(E)-3-(dimethylamino)prop-2-enoyl]phenyl]-N-ethylacetamide
Molekylvikt (g/mol) 260.33
Kvantitet 4 x 25 mg
Formel vikt 260.1525 g/mol