missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,4-Difluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60452.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496918

  • 6855.28 SEK / 1g


Estimated Shipment date
from Supplier 15-07-2019
Add to basket

Specifications

Chemical Identifiers

330468-44-5
269.27
UKFHAYWBZJDMKO-UHFFFAOYSA-N
798868
C12H9F2NO2S
MFCD01213712
n-3,4-difluorophenyl benzenesulfonamide, 3,4-difluorophenyl phenylsulfonyl amine, n-3,4-difluoro-phenyl-benzenesulfonamide, n-3,4-difluorophenyl-1-benzenesulfonamide

Specifications

N-(3,4-Difluorophenyl)benzenesulfonamide
330468-44-5
MFCD01213712
UKFHAYWBZJDMKO-UHFFFAOYSA-N
798868
Solid
1g
C12H9F2NO2S
n-3,4-difluorophenyl benzenesulfonamide, 3,4-difluorophenyl phenylsulfonyl amine, n-3,4-difluoro-phenyl-benzenesulfonamide, n-3,4-difluorophenyl-1-benzenesulfonamide
269.27
269.27
97%
Videos
SDS