Läs mer
N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole Sulfide(Mixture of 1 and 3 isomers), TRC
Produktkod. 30374350
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374350 250 mg
30376013 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30374350 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD470965250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 957470-58-5
Molekylformel C24H24F2N4O5S
Molekylvikt (g/mol) 518.53
Synonym 5-(Difluoromethoxy)-1-[(3,4-dimethoxy-2-pyridinyl)methyl]-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]thio]-1H-benzimidazole
LEDER COc1ccnc(CSc2nc3cc(OC(F)F)ccc3n2Cc4nccc(OC)c4OC)c1OC
Specifikationer
Kemiskt namn eller material N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole Sulfide(Mixture of 1 and 3 isomers)
Kvantitet 250 mg
InChI formel InChI=1S/C24H24F2N4O5S/c1-31-19-7-9-27-16(21(19)33-3)12-30-18-6-5-14(35-23(25)26)11-15(18)29-24(30)36-13-17-22(34-4)20(32-2)8-10-28-17/h5-11,23H,12-13H2,1-4H3
Formel vikt 518.1435