Läs mer
N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole Sulfone(Mixture of 1 and 3 isomers), TRC
Produktkod. 30375004
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375004 10 mg
30379871 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30375004 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47097010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797132-60-5
Molekylformel C24H24F2N4O7S
Molekylvikt (g/mol) 550.53
Synonym 5-(Difluoromethoxy)-1-[(3,4-dimethoxy-2-pyridinyl)methyl]-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole, Pantoprazole Impurity
LEDER COc1ccnc(Cn2c(nc3cc(OC(F)F)ccc23)S(=O)(=O)Cc4nccc(OC)c4OC)c1OC
Specifikationer
Kemiskt namn eller material N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole Sulfone(Mixture of 1 and 3 isomers)
Kvantitet 10 mg
InChI formel InChI=1S/C24H24F2N4O7S/c1-33-19-7-9-27-16(21(19)35-3)12-30-18-6-5-14(37-23(25)26)11-15(18)29-24(30)38(31,32)13-17-22(36-4)20(34-2)8-10-28-17/h5-11,23H,12-13H2,1-4H3
Formel vikt 550.1334