missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,5-Difluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65926.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450017

  • 2716.64 SEK / 1g


Estimated Shipment date
from Supplier 17-12-2018
Add to basket

Specifications

Chemical Identifiers

MFCD28098842

Specifications

N-(3,5-Difluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
MFCD28098842
Solid
1g
287.31
95%
Videos
SDS