missing translation for 'onlineSavingsMsg'
Läs mer

N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetamide Hydrochloride (Diacetolol Hydrochloride), Mikromol™

Produktkod. 30052136
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052136 25 mg
18042900 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30052136 Leverantör Mikromol Leverantörsnummer MM0435.020025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetamide Hydrochloride (Diacetolol Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Diacetolol Hydrochloride
CAS 73899-76-0
Analyt- eller komponentnamn Diacetolol Hydrochloride
Frakt skick Room Temperature
Molekylformel C16 H24 N2 O4 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Acebutolol Hydrochloride Imp. B (EP), Acebutolol Imp. B (EP), Acebutolol USP Related Compound B, Acebutolol USP RC B, N-[3-Acetyl-4-[(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetamide Hydrochloride, Acebutolol Hydrochloride Imp. B (EP) as Hydrochloride, Diacetolol Hydrochloride, Acebutolol Related Compound B as Hydrochloride, Acebutolol Hydrochloride Impurity B as Hydrochloride, Acebutolol Impurity B as Hydrochloride
InChI formel InChI=1S/C16H24N2O4.ClH/c1-10(2)17-8-14(21)9-22-16-6-5-13(18-12(4)20)7-15(16)11(3)19;/h5-7,10,14,17,21H,8-9H2,1-4H3,(H,18,20);1H
LEDER Cl.CC(C)NCC(O)COc1ccc(NC(=O)C)cc1C(=O)C
IUPAC-namn N-[3-acetyl-4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide;hydrochloride
Molekylvikt (g/mol) 344.83
Kvantitet 25 mg
Formel vikt 344.1503 g/mol
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.