missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Bromophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59013.03

Additional Details : CAS Number : 36293-19-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465178

  • 5659.56 SEK / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Bromophenyl)-3,5-dinitrobenzamide
366.12
2895320
36293-19-3
MFCD01010633

Specifications

N-(3-Bromophenyl)-3,5-dinitrobenzamide
1g
97%
366.12
2895320
36293-19-3
Solid
Off-white
MFCD01010633
Videos
SDS