Läs mer
N-(3-Carboxy-1-oxopropyl)glycylglycyl-N-(4-nitrophenyl)-L-phenylalaninamide, TRC
Produktkod. 30353460
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353460 10 mg
30354175 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43286010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68982-90-1
Molekylformel C23H25N5O8
Molekylvikt (g/mol) 499.47
Synonym (S)-4-((2-((2-((1-((4-Nitrophenyl)amino)-1-oxo-3-phenylpropan-2-yl)amino)-2-oxoethyl)amino)-2-oxoethyl)amino)-4-oxobutanoic Acid, Chymotrypsin Substrate I, Colorimetric
IUPAC-namn 4-[[2-[[2-[[(2S)-1-(4-nitroanilino)-1-oxo-3-phenylpropan-2-yl]amino]-2-oxoethyl]amino]-2-oxoethyl]amino]-4-oxobutanoic acid
LEDER OC(=O)CCC(=O)NCC(=O)NCC(=O)N[C@@H](Cc1ccccc1)C(=O)Nc2ccc(cc2)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material N-(3-Carboxy-1-oxopropyl)glycylglycyl-N-(4-nitrophenyl)-L-phenylalaninamide
Kvantitet 10 mg
InChI formel InChI=1S/C23H25N5O8/c29-19(10-11-22(32)33)24-13-20(30)25-14-21(31)27-18(12-15-4-2-1-3-5-15)23(34)26-16-6-8-17(9-7-16)28(35)36/h1-9,18H,10-14H2,(H,24,29)(H,25,30)(H,26,34)(H,27,31)(H,32,33)/t18-/m0/s1
Formel vikt 499.1703