missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chloro-4-methoxyphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65535.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489716

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chloro-4-methoxyphenyl)-2,2-diphenylacetamide
MFCD01060914

Specifications

N-(3-Chloro-4-methoxyphenyl)-2,2-diphenylacetamide
Solid
MFCD01060914
351.83
95%
1g
Videos
SDS