missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chlorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60471.03

Additional Details : CAS Number : 54129-19-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466938

  • 5659.56 SEK / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chlorophenyl)benzenesulfonamide
54129-19-0
267.73
MFCD00552411

Specifications

N-(3-Chlorophenyl)benzenesulfonamide
Solid
267.73
Off-white
54129-19-0
97%
MFCD00552411
1g
Videos
SDS