Läs mer
N-[3-Fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]carbamic Acid Phenylmethyl Ester, TRC
Produktkod. 30408384
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408384 10 mg
30395215 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30408384 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59363510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[3-Fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]carbamic Acid Phenylmethyl Ester
CAS 1220910-89-3
Molekylformel C21H17FN6O2
Kvantitet 10 mg
Synonym Tedizolid Impurity 68
InChI formel InChI=1S/C21H17FN6O2/c1-28-26-20(25-27-28)19-10-7-15(12-23-19)17-9-8-16(11-18(17)22)24-21(29)30-13-14-5-3-2-4-6-14/h2-12H,13H2,1H3,(H,24,29)
LEDER FC1=C(C2=CC=C(C3=NN(C)N=N3)N=C2)C=CC(NC(OCC4=CC=CC=C4)=O)=C1
IUPAC-namn benzyl (3-fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)carbamate
Molekylvikt (g/mol) 404.4
Formel vikt 404.14