Learn More
Alfa Aesar™ N-(3-Fluoro-5-methylphenyl)-2,2-diphenylacetamide, 95%
Brand:  Alfa Aesar™ H65692.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15499836
  • 2716.64 SEK / 1g
Estimated Shipment date
from Supplier 16-07-2019
Add to basket
Specifications
Chemical Identifiers
MFCD27975806
Specifications
N-(3-Fluoro-5-methylphenyl)-2,2-diphenylacetamide
MFCD27975806
Solid
1g
319.38
95%