Learn More
Alfa Aesar™ N-(3-Fluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%
Brand:  Alfa Aesar™ H65091.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469366
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 31-07-2018
Add to basket
Specifications
Chemical Identifiers
N-(3-Fluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Specifications
N-(3-Fluorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
1g
269.32
95%