Läs mer
N-(3-Indolylacetyl)glycine, TRC
Produktkod. 30012101
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012101 2.5 g 2.5g
30011571 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI6280052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13113-08-1
Molekylformel C12 H12 N2 O3
Molekylvikt (g/mol) 232.24
Synonym N-[2-(1H-Indol-3-yl)acetyl]glycine, N-(1H-Indol-3-ylacetyl)glycine, Indole-3-acetylglycine
IUPAC-namn 2-[[2-(1H-indol-3-yl)acetyl]amino]acetic acid
LEDER OC(=O)CNC(=O)Cc1c[nH]c2ccccc12
Specifikationer
Kemiskt namn eller material N-(3-Indolylacetyl)glycine
InChI formel InChI=1S/C12H12N2O3/c15-11(14-7-12(16)17)5-8-6-13-10-4-2-1-3-9(8)10/h1-4,6,13H,5,7H2,(H,14,15)(H,16,17)
Kvantitet 2.5 g
Formel vikt 232.0848