missing translation for 'onlineSavingsMsg'
Läs mer

N-[3-Methyl-1-(1-phenylcyclobutyl)butyl]-N,N-dimethylamine Hydrochloride, TRC

Produktkod. 30470375
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30470375 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470375 Supplier Toronto Research Chemicals Supplier No. TRCM326058100MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-[3-Methyl-1-(1-phenylcyclobutyl)butyl]-N,N-dimethylamine hydrochloride
CAS 84485-01-8
Molekylformel C17 H27 N . Cl H
Kvantitet 100 mg
Synonym N,N-Dimethyl-α-(2-methylpropyl)-1-phenylcyclobutanemethanamine Hydrochloride (1:1), N,N,3-Trimethyl-1-(1-phenylcyclobutyl)butan-1-amine hydrochloride, N-[3-Methyl-1-(1-phenylcyclobutyl)butyl]-N,N-dimethylamine hydrochloride
InChI formel InChI=1S/C17H27N.ClH/c1-14(2)13-16(18(3)4)17(11-8-12-17)15-9-6-5-7-10-15;/h5-7,9-10,14,16H,8,11-13H2,1-4H3;1H
LEDER Cl.CC(C)CC(N(C)C)C1(CCC1)c2ccccc2
IUPAC-namn N,N,3-trimethyl-1-(1-phenylcyclobutyl)butan-1-amine;hydrochloride
Molekylvikt (g/mol) 281.86
Formel vikt 281.191
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.