Läs mer
N-[3-(Methylthio)-1-oxopropyl]glycine, TRC
Produktkod. 30479051
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479051 50 mg
30464645 250 mg
30476948 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479051 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM17416050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96305-44-1
Molekylformel C6 H11 N O3 S
Molekylvikt (g/mol) 177.221
Synonym 2-[3-(Methylsulfanyl)propanamido]acetic Acid, (3-(Methylthio)propanoyl)glycine
IUPAC-namn 2-(3-methylsulfanylpropanoylamino)acetic acid
LEDER CSCCC(=O)NCC(=O)O
Specifikationer
Kemiskt namn eller material N-[3-(Methylthio)-1-oxopropyl]glycine
Kvantitet 50 mg
InChI formel InChI=1S/C6H11NO3S/c1-11-3-2-5(8)7-4-6(9)10/h2-4H2,1H3,(H,7,8)(H,9,10)
Formel vikt 177.046