Läs mer
N-(3-Morpholinopropyl) Gefitinib, TRC
Produktkod. 30136572
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136572 100 mg
17981235 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136572 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM725210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1502829-45-9
Molekylformel C29 H37 Cl F N5 O4
Molekylvikt (g/mol) 574.087
Synonym 4-Quinazolinamine, N-(3-chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-N-[3-(4-morpholinyl)propyl]-, N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-N-[3-(4-morpholinyl)propyl]-4-quinazolinamine, N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine
IUPAC-namn N-(3-chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholin-4-ylpropoxy)-N-(3-morpholin-4-ylpropyl)quinazolin-4-amine
LEDER COc1cc2ncnc(N(CCCN3CCOCC3)c4ccc(F)c(Cl)c4)c2cc1OCCCN5CCOCC5
Specifikationer
Kemiskt namn eller material N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine
Kvantitet 100 mg
InChI formel InChI=1S/C29H37ClFN5O4/c1-37-27-20-26-23(19-28(27)40-13-3-7-35-11-16-39-17-12-35)29(33-21-32-26)36(22-4-5-25(31)24(30)18-22)8-2-6-34-9-14-38-15-10-34/h4-5,18-21H,2-3,6-17H2,1H3
Formel vikt 573.252