Learn More
Alfa Aesar™ N-[4-(2-Thiazolidinyl)phenyl]acetamide, 97%
Brand:  Alfa Aesar™ H62010.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15458608
  • 10374.25 SEK / 5g
Estimated Shipment date
from Supplier 03-08-2018
Add to basket
Specifications
Chemical Identifiers
N-[4-(2-Thiazolidinyl)phenyl]acetamide
MFCD09027147
2-(4-Acetamidophenyl)thiazolidine
Specifications
N-[4-(2-Thiazolidinyl)phenyl]acetamide
5g
97%
2-(4-Acetamidophenyl)thiazolidine
222.31
Solid
MFCD09027147