Läs mer
N-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl] Rabeprazole Sulfide, TRC
Produktkod. 30471595
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471595 500 mg
30466364 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30471595 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM265175500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1114543-47-3
Molekylformel C29 H36 N4 O4 S
Molekylvikt (g/mol) 536.69
Synonym 1-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]-2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole, Rabeprazole N-Alkyl Sulfide
IUPAC-namn 1-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl]-2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfanyl]benzimidazole
LEDER COCCCOc1ccnc(CSc2nc3ccccc3n2Cc4nccc(OCCCOC)c4C)c1C
Specifikationer
Kemiskt namn eller material N-[(4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl)methyl] Rabeprazole Sulfide
Kvantitet 500 mg
InChI formel InChI=1S/C29H36N4O4S/c1-21-24(30-13-11-27(21)36-17-7-15-34-3)19-33-26-10-6-5-9-23(26)32-29(33)38-20-25-22(2)28(12-14-31-25)37-18-8-16-35-4/h5-6,9-14H,7-8,15-20H2,1-4H3
Formel vikt 536.2457