Learn More
N-(4,6-Dimethyl-2-pyridyl)thiourea, 97%, Thermo Scientific™
Product Code. 15496507
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496507 5 g 5g
15406517 250 mg 250mg
15486507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15496507 Supplier Thermo Scientific Alfa Aesar Supplier No. H33661.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 49600-35-3
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.26
MDL Number MFCD15525456
InChI Key ZXQTZSMIBLLDGW-UHFFFAOYSA-N
Synonym 4,6-dimethyl-pyridin-2-yl-thiourea, n-4,6-dimethyl-2-pyridinyl thiourea, n-4,6-dimethyl-2-pyridyl thiourea
PubChem CID 13608509
IUPAC Name (4,6-dimethylpyridin-2-yl)thiourea
SMILES CC1=CC(C)=NC(NC(N)=S)=C1
Specifications
Melting Point 238°C to 243°C
Quantity 5 g
Formula Weight 181.25
Percent Purity 97%
Chemical Name or Material N-(4,6-Dimethyl-2-pyridyl)thiourea