Läs mer
N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide (Diacetylsulfadimidine; Diacetylsulfamethazine), Mikromol™
Produktkod. 30052654
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052654 25 mg
30049084 100 mg
30460756 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30052654 Leverantör Mikromol Leverantörsnummer MM1311.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide (Diacetylsulfadimidine; Diacetylsulfamethazine)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
CAS 59224-69-0
Analyt- eller komponentnamn N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
Frakt skick Room Temperature
Molekylformel C16 H18 N4 O4 S
Certifikat och efterlevnad ISO 17025
Synonym N-[[4-(Acetylamino)phenyl]sulfonyl]-N-(4,6-dimethyl-2-pyrimidinyl)acetamide
InChI formel InChI=1S/C16H18N4O4S/c1-10-9-11(2)18-16(17-10)20(13(4)22)25(23,24)15-7-5-14(6-8-15)19-12(3)21/h5-9H,1-4H3,(H,19,21)
LEDER CC(=O)Nc1ccc(cc1)S(=O)(=O)N(C(=O)C)c2nc(C)cc(C)n2
IUPAC-namn N-[4-[acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
Molekylvikt (g/mol) 362.4
Kvantitet 25 mg
Formel vikt 362.1049 g/mol