Läs mer
N-(4-Amino-6,7-dimethoxyquinazol-2-yl)-N-methylpropylenediamine Formamide Hydrochloride, TRC
Produktkod. 30137147
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137147 100 mg
30006298 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA604975100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1796934-51-4
Molekylformel C15H21N5O3 . HCl
Molekylvikt (g/mol) 355.82
Synonym N[3-[(4-Amino-6,7-dimethoxyquinaolin-2-yl)methylamino]propyl]formamide Hydrochloride, Alfuzosin Hydrochloride Impurity E (EP)
IUPAC-namn N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]formamide;hydrochloride
LEDER Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC=O
Specifikationer
Kemiskt namn eller material N-(4-Amino-6,7-dimethoxyquinazol-2-yl)-N-methylpropylenediamine Formamide Hydrochloride
Analyt- eller komponentnamn N-(4-Amino-6,7-dimethoxyquinazol-2-yl)-N-methylpropylenediamine Formamide Hydrochloride
Frakt skick Room Temperature
InChI formel InChI=1S/C15H21N5O3.ClH/c1-20(6-4-5-17-9-21)15-18-11-8-13(23-3)12(22-2)7-10(11)14(16)19-15;/h7-9H,4-6H2,1-3H3,(H,17,21)(H2,16,18,19);1H
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 100 mg
Formel vikt 355.1411