Learn More
N-(4-Aminobenzoyl)-L-glutamic Acid, TRC
Product Code. 17987734
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987734 1 g 1g
30005415 5 g 5g
17908814 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17987734 Supplier Toronto Research Chemicals Supplier No. TRCA5937701G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4271-30-1
Molecular Formula C12 H14 N2 O5
Molecular Weight (g/mol) 266.25
Synonym (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid, Folic Acid Imp. A (EP), N-(4-Aminobenzoyl)-L-glutamic Acid, Glutamic acid, N-(p-aminobenzoyl)- (6CI,7CI), Glutamic acid, N-(p-aminobenzoyl)-, L- (8CI), (4-Aminobenzoyl)-L-glutamic acid, (p-Aminobenzoyl)-L-glutamic acid, (p-Aminobenzoyl)glutamic acid, N-(p-Aminobenzoyl)-L(+)-glutamic acid, N-(p-Aminobenzoyl)-L-glutamic acid, N-(p-Aminobenzoyl)glutamic acid, NSC 71042
IUPAC Name (2S)-2-[(4-aminobenzoyl)amino]pentanedioic acid
SMILES Nc1ccc(cc1)C(=O)N[C@@H](CCC(=O)O)C(=O)O
Specifications
Analyte or Component Names (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C12H14N2O5/c13-8-3-1-7(2-4-8)11(17)14-9(12(18)19)5-6-10(15)16/h1-4,9H,5-6,13H2,(H,14,17)(H,15,16)(H,18,19)/t9-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Formula Weight 266.0903
Chemical Name or Material (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)