Learn More
N-4-Benzyloxyphenyl Alpha-Benzilidene-d5 Isobutyrylacetamide, TRC
Product Code. 30333247
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30333247 50 mg 50mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30333247 Supplier Toronto Research Chemicals Supplier No. TRCB28763350MG
 Restricted Item
Beräknad leverans: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-4-Benzyloxyphenyl a-Benzilidene-d5 Isobutyrylacetamide
CAS 1020719-20-3
Molecular Formula C26 D5 H20 N O3
Quantity 50 mg
Synonym 4-Methyl-3-oxo-N-[4-(phenylmethoxy)phenyl]-2-(phenyl-2,3,4,5,6-d5-methylene)pentanamimde
InChI Formula InChI=1S/C26H25NO3/c1-19(2)25(28)24(17-20-9-5-3-6-10-20)26(29)27-22-13-15-23(16-14-22)30-18-21-11-7-4-8-12-21/h3-17,19H,18H2,1-2H3,(H,27,29)/b24-17+/i3D,5D,6D,9D,10D
SMILES [2H]c1c([2H])c([2H])c(\C=C(\C(=O)Nc2ccc(OCc3ccccc3)cc2)/C(=O)C(C)C)c([2H])c1[2H]
IUPAC Name (2E)-4-methyl-3-oxo-2-[(2,3,4,5,6-pentadeuteriophenyl)methylidene]-N-(4-phenylmethoxyphenyl)pentanamide
Molecular Weight (g/mol) 404.51
Formula Weight 404.2148