Learn More
Alfa Aesar™ N-(4-Bromo-2-chlorophenyl)-4-chloro-3-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65295.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449546
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 12-06-2018
Add to basket
Specifications
Chemical Identifiers
N-(4-Bromo-2-chlorophenyl)-4-chloro-3-sulfamoylbenzamide
424.1
MFCD28098780
Specifications
N-(4-Bromo-2-chlorophenyl)-4-chloro-3-sulfamoylbenzamide
95%
MFCD28098780
Solid
424.1
1g