Learn More
Alfa Aesar™ N-(4-Bromo-2-chlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%
Brand:  Alfa Aesar™ H65264.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449516
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 12-03-2018
Add to basket
Specifications
Chemical Identifiers
N-(4-Bromo-2-chlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
364.67
MFCD28098775
Specifications
N-(4-Bromo-2-chlorophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
95%
MFCD28098775
Solid
364.67
1g