missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-methylphenyl)-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65378.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499596

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-methylphenyl)-4-chloro-3-sulfamoylbenzamide
403.68
MFCD27975754

Specifications

N-(4-Bromo-2-methylphenyl)-4-chloro-3-sulfamoylbenzamide
95%
MFCD27975754
Solid
403.68
1g
Videos
SDS