Learn More
Alfa Aesar™ N-(4-Bromo-3-fluorophenyl)-2,2-diphenylacetamide, 95%
Brand:  Alfa Aesar™ H65245.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449496
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 03-09-2018
Add to basket
Specifications
Chemical Identifiers
N-(4-Bromo-3-fluorophenyl)-2,2-diphenylacetamide
MFCD27975742
Specifications
N-(4-Bromo-3-fluorophenyl)-2,2-diphenylacetamide
MFCD27975742
384.25
95%
1g
Solid