Learn More
Alfa Aesar™ N-(4-Bromo-3-methylphenyl)-2,2-diphenylacetamide, 95%
Brand:  Alfa Aesar™ H65610.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15459776
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Chemical Identifiers
N-(4-Bromo-3-methylphenyl)-2,2-diphenylacetamide
380.29
MFCD03241519
Specifications
N-(4-Bromo-3-methylphenyl)-2,2-diphenylacetamide
95%
MFCD03241519
Solid
380.29
1g