Läs mer
N-(4-Bromo-3-methylphenyl)-carbamic Acid Phenylmethyl Ester, TRC
Produktkod. 30342222
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30342222 500 mg
1 options
Produktkod. 30342222 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB699873500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(4-Bromo-3-methylphenyl)-carbamic Acid Phenylmethyl Ester
CAS 1246187-55-2
Molekylformel C15H14BrNO2
Kvantitet 500 mg
Synonym Benzyl (4-bromo-3-methylphenyl)carbamate
InChI formel InChI=1S/C15H14BrNO2/c1-11-9-13(7-8-14(11)16)17-15(18)19-10-12-5-3-2-4-6-12/h2-9H,10H2,1H3,(H,17,18)
LEDER Cc1cc(NC(=O)OCc2ccccc2)ccc1Br
IUPAC-namn benzyl N-(4-bromo-3-methylphenyl)carbamate
Molekylvikt (g/mol) 320.18
Formel vikt 319.0208