missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chloro-2-fluorophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62684.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497726

  • 13699.94 SEK / 5g


Estimated Shipment date
from Supplier 25-07-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26143334

Specifications

N-(4-Chloro-2-fluorophenyl)-3,5-difluorobenzenesulfonamide
MFCD26143334
Solid
5g
321.7
97%
Videos
SDS