Läs mer
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide, TRC
Produktkod. 30366364
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366364 5 g
30366350 1 g
30354269 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3650135G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 848133-76-6
Molekylformel C14H12ClN3O2
Molekylvikt (g/mol) 289.72
Synonym 4-Chloro-3-cyano-7-ethoxy-6-(N-acetylamino)quinoline, 4-Chloro-3-cyano-7-ethoxy-6-(acetylamino)quinoline, N-(4-Chloro-3-cyano-7-ethoxyquinolin-6-yl)acetamide
IUPAC-namn N-(4-chloro-3-cyano-7-ethoxyquinolin-6-yl)acetamide
LEDER CCOc1cc2ncc(C#N)c(Cl)c2cc1NC(=O)C
Specifikationer
Kemiskt namn eller material N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide
Kvantitet 5 g
InChI formel InChI=1S/C14H12ClN3O2/c1-3-20-13-5-11-10(4-12(13)18-8(2)19)14(15)9(6-16)7-17-11/h4-5,7H,3H2,1-2H3,(H,18,19)