Läs mer
N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, TRC
Produktkod. 30179748
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179748 100 mg
30180360 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179748 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC380130100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2457316-06-0
Molekylformel C16 H15 Cl N2 O3 S
Molekylvikt (g/mol) 350.82
Synonym N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, Benzenesulfonamide, 2-chloro-5-[(2,3-dihydro-2-methyl-1H-indol-1-yl)carbonyl]- (ACI), 2-Chloro-5-[(2,3-dihydro-2-methyl-1H-indol-1-yl)carbonyl]benzenesulfonamide (ACI)
IUPAC-namn 2-chloro-5-(2-methyl-2,3-dihydroindole-1-carbonyl)benzenesulfonamide
LEDER CC1Cc2ccccc2N1C(=O)c3ccc(Cl)c(c3)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline
Kvantitet 100 mg
InChI formel InChI=1S/C16H15ClN2O3S/c1-10-8-11-4-2-3-5-14(11)19(10)16(20)12-6-7-13(17)15(9-12)23(18,21)22/h2-7,9-10H,8H2,1H3,(H2,18,21,22)
Formel vikt 350.0492