missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chlorobenzyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59837.03

Additional Details : CAS Number : 304645-25-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496258

  • 5659.56 SEK / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chlorobenzyl)-2-fluorobenzamide
304645-25-8
263.7
MFCD01339839

Specifications

N-(4-Chlorobenzyl)-2-fluorobenzamide
Solid
263.7
1g
304645-25-8
97%
MFCD01339839
Videos
SDS