missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chlorophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61870.03

Additional Details : CAS Number : 948710-52-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488458

  • 5659.56 SEK / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chlorophenyl)-3,5-difluorobenzamide
267.66
15747071
948710-52-9
MFCD04806351

Specifications

N-(4-Chlorophenyl)-3,5-difluorobenzamide
Solid
267.66
15747071
1g
948710-52-9
97%
MFCD04806351
White
Videos
SDS