Läs mer
N-(4'-Cyano[1,1'-biphenyl]-4-yl)-N'-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl] Urea, TRC
Produktkod. 30359073
Change view
Kvantitet:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359073 25 mg
30355391 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359073 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC96234525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1361414-26-7
Molekylformel C26H18N6O
Molekylvikt (g/mol) 430.46
Synonym SR-9186
IUPAC-namn 1-[4-(4-cyanophenyl)phenyl]-3-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl]urea
LEDER O=C(NC1=CC=C(C2=CC=C(C#N)C=C2)C=C1)NC(C=C3)=CC=C3C4=C5C(NC=N5)=NC=C4
Specifikationer
Kemiskt namn eller material N-(4'-Cyano[1,1'-biphenyl]-4-yl)-N'-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl] Urea
Kvantitet 25 mg
InChI formel InChI=1S/C26H18N6O/c27-15-17-1-3-18(4-2-17)19-5-9-21(10-6-19)31-26(33)32-22-11-7-20(8-12-22)23-13-14-28-25-24(23)29-16-30-25/h1-14,16H,(H,28,29,30)(H2,31,32,33)
Formel vikt 430.1542