Läs mer
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide, TRC
Produktkod. 30363169
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363169 100 mg
30360979 50 mg
30362533 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC778030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 316373-92-9
Molekylformel C12H11F3N2O3
Molekylvikt (g/mol) 288.22
Synonym Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methyl-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide
IUPAC-namn N-[4-cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide
LEDER CC(O)(CO)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F
Specifikationer
Kemiskt namn eller material N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide
Kvantitet 100 mg
InChI formel InChI=1S/C12H11F3N2O3/c1-11(20,6-18)10(19)17-8-3-2-7(5-16)9(4-8)12(13,14)15/h2-4,18,20H,6H2,1H3,(H,17,19)
Formel vikt 288.0722