Läs mer
N-(4-Difluoromethoxy-2-nitrophenyl)acetamide, TRC
Produktkod. 30368001
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368001 50 mg
30362191 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368001 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD36709050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97963-75-2
Molekylformel C9H8F2N2O4
Molekylvikt (g/mol) 246.17
Synonym N-[4-(Difluoromethoxy)-2-nitrophenyl]acetamide, Pantoprazole Impurity 9
IUPAC-namn N-[4-(difluoromethoxy)-2-nitrophenyl]acetamide
LEDER CC(=O)Nc1ccc(OC(F)F)cc1[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material N-(4-Difluoromethoxy-2-nitrophenyl)acetamide
Kvantitet 50 mg
InChI formel InChI=1S/C9H8F2N2O4/c1-5(14)12-7-3-2-6(17-9(10)11)4-8(7)13(15)16/h2-4,9H,1H3,(H,12,14)
Formel vikt 246.0452