missing translation for 'onlineSavingsMsg'
Läs mer

N-(4-Ethoxyphenyl)maleamic acid, 97%, Thermo Scientific™

Produktkod. p-7027948
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11425987 5 g 5g
11435987 25 g 25g
11445987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11425987 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B25024.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 108087-84-9
Molekylformel C12H13NO4
Molekylvikt (g/mol) 235.239
MDL-nummer MFCD00020477
InChI-nyckel LMTIUWRMRAZPEN-FPLPWBNLSA-N
Synonym z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid, 2z-4-4-ethoxyphenyl amino-4-oxobut-2-enoic acid, n-4-ethoxyphenyl maleamic acid, 2z-3-n-4-ethoxyphenyl carbamoyl prop-2-enoic acid, 2z-3-4-ethoxyphenyl carbamoyl prop-2-enoic acid, 4-4-ethoxyanilino-4-oxobut-2-enoic acid, z-4-4-ethoxyanilino-4-oxobut-2-enoic acid, 2-butenoic acid,4-4-ethoxyphenyl amino-4-oxo-, 2z
PubChem CID 1808883
IUPAC-namn (Z)-4-(4-ethoxyanilino)-4-oxobut-2-enoic acid
LEDER CCOC1=CC=C(C=C1)NC(=O)C=CC(=O)O

Specifikationer

Kemiskt namn eller material N-(4-Ethoxyphenyl)maleamic acid
Smältpunkt 188°C to 189°C
Kvantitet 5 g
Formel vikt 235.24
Procent renhet 97%
Produkttitel
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.