Läs mer
N-[(4-Hydroxy-3-methoxyphenyl)methyl]octanamide (N-Vanillyloctanamide), Mikromol™
Produktkod. 17931826
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17931826 100 mg
17940786 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17931826 Leverantör Mikromol Leverantörsnummer MM1033.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[(4-Hydroxy-3-methoxyphenyl)methyl]octanamide (N-Vanillyloctanamide)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[(4-Hydroxy-3-methoxyphenyl)methyl]-octanamide (N-Vanillyloctanamide)
CAS 58493-47-3
Analyt- eller komponentnamn N-[(4-Hydroxy-3-methoxyphenyl)methyl]-octanamide (N-Vanillyloctanamide)
Frakt skick Room Temperature
Molekylformel C16 H25 N O3
Certifikat och efterlevnad ISO 17025
Synonym N-[(4-Hydroxy-3-methoxyphenyl)methyl]-octanamide, N-Vanillyloctanamide
InChI formel InChI=1S/C16H25NO3/c1-3-4-5-6-7-8-16(19)17-12-13-9-10-14(18)15(11-13)20-2/h9-11,18H,3-8,12H2,1-2H3,(H,17,19)
LEDER CCCCCCCC(=O)NCc1ccc(O)c(OC)c1
IUPAC-namn N-[(4-hydroxy-3-methoxyphenyl)methyl]octanamide
Molekylvikt (g/mol) 279.37
Kvantitet 100 mg
Formel vikt 279.1834 g/mol