Läs mer
N-(4-Hydroxyphenyl)-5-methyl-2-1H-pyridone, TRC
Produktkod. 30407852
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407852 50 mg
30404025 10 mg
30405291 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30407852 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94940050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 851518-71-3
Molekylformel C12 H11 N O2
Molekylvikt (g/mol) 201.22
Synonym 2(1H)-Pyridinone, 1-(4-hydroxyphenyl)-5-methyl-, 1-(4-Hydroxyphenyl)-5-methyl-2(1H)-pyridinone, F 351, N-(4-Hydroxyphenyl)-5-methyl-2-1H-pyridone, 4'-Hydroxypirfenidone
IUPAC-namn 1-(4-hydroxyphenyl)-5-methylpyridin-2-one
LEDER CC1=CN(C(=O)C=C1)c2ccc(O)cc2
Specifikationer
Kemiskt namn eller material N-(4-Hydroxyphenyl)-5-methyl-2-1H-pyridone
Kvantitet 50 mg
InChI formel InChI=1S/C12H11NO2/c1-9-2-7-12(15)13(8-9)10-3-5-11(14)6-4-10/h2-8,14H,1H3
Formel vikt 201.079