missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Methoxybenzyl)-N-methylamine, tech, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Density | 1.0080g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3C6H4CH2NCH3 |
Quantity | 250 mg |
Formula Weight | 151.21 |
Percent Purity | 97% |
Chemical Name or Material | N-(4-Methoxybenzyl)-N-methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'