Läs mer
N-4-Methoxyphenylretinamide, TRC
Produktkod. 30468874
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468874 50 mg
30482646 5 mg
30478467 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30468874 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26266050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79965-10-9
Molekylformel C27 H35 N O2
Molekylvikt (g/mol) 405.57
Synonym 4-Methoxy Fenretinide
IUPAC-namn (2E,4E,6E,8E)-N-(4-methoxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide
LEDER COc1ccc(NC(=O)\C=C(/C)\C=C\C=C(/C)\C=C\C2=C(C)CCCC2(C)C)cc1
Specifikationer
Kemiskt namn eller material N-4-Methoxyphenylretinamide
Kvantitet 50 mg
InChI formel InChI=1S/C27H35NO2/c1-20(12-17-25-22(3)11-8-18-27(25,4)5)9-7-10-21(2)19-26(29)28-23-13-15-24(30-6)16-14-23/h7,9-10,12-17,19H,8,11,18H2,1-6H3,(H,28,29)/b10-7+,17-12+,20-9+,21-19+
Formel vikt 405.2668