Läs mer
N-[4-Methyl-3-(4-pyridin-3-yl-pyrimidin-2-ylamino)-phenyl]-4-chloromethyl Benzamide, TRC
Produktkod. 30474353
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474353 10 mg
30481626 50 mg
30477649 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32594310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 404844-11-7
Molekylformel C24 H20 Cl N5 O
Molekylvikt (g/mol) 429.9
Synonym Benzamide, 4-(chloromethyl)-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, 2-Methyl-5-[[4-(chloromethyl)benzoyl]amino]-N-[4-(pyridin-3-yl)pyrimidin-2-yl]benzenamine, 4-(Chloromethyl)-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, 4-Chloromethyl-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide, N-[5-[[4-(Chloromethyl)benzoyl]amino]-2-methylphenyl]-4-(3-pyridyl)-2-pyrimidineamine
IUPAC-namn 4-(chloromethyl)-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
LEDER Cc1ccc(NC(=O)c2ccc(CCl)cc2)cc1Nc3nccc(n3)c4cccnc4
Specifikationer
Kemiskt namn eller material 4-(Chloromethyl)-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide
Kvantitet 10 mg
InChI formel InChI=1S/C24H20ClN5O/c1-16-4-9-20(28-23(31)18-7-5-17(14-25)6-8-18)13-22(16)30-24-27-12-10-21(29-24)19-3-2-11-26-15-19/h2-13,15H,14H2,1H3,(H,28,31)(H,27,29,30)
Formel vikt 429.1356