Läs mer
N-[4-(Methylamino)-3-nitrobenzoyl]-N-2-pyridinyl-Beta-alanine-d3 Ethyl Ester Hydrochloride, TRC
Produktkod. 30468096
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30468096 100 mg
1 options
Produktkod. 30468096 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM286582100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[4-(Methylamino)-3-nitrobenzoyl]-N-2-pyridinyl-beta-alanine-d3 Ethyl Ester Hydrochloride
CAS 2124271-66-3
Molekylformel C18H18D3ClN4O5
Kvantitet 100 mg
Synonym Ethyl 3-[[4-(Methylamino)-3-nitrobenzoyl](pyridin-2-yl)amino]propanoate-d3 Hydrochloride
InChI formel InChI=1S/C18H20N4O5.ClH/c1-3-27-17(23)9-11-21(16-6-4-5-10-20-16)18(24)13-7-8-14(19-2)15(12-13)22(25)26;/h4-8,10,12,19H,3,9,11H2,1-2H3;1H/i2D3;
LEDER Cl.[2H]C([2H])([2H])Nc1ccc(cc1[N+](=O)[O-])C(=O)N(CCC(=O)OCC)c2ccccn2
IUPAC-namn ethyl 3-[[3-nitro-4-(trideuteriomethylamino)benzoyl]-pyridin-2-ylamino]propanoate;hydrochloride
Molekylvikt (g/mol) 411.85
Formel vikt 411.1389