missing translation for 'onlineSavingsMsg'
Learn More

N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide, Mikromol™

Product Code. 30052476
Change view
Click to view available options
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30052476 25 mg
18004010 100 mg
30459242 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052476 Supplier Mikromol Supplier No. MM0161.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide
CAS 13312-12-4
Analyt- eller komponentnamn N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide
Frakt skick Room Temperature
Molekylformel C10 H9 F3 N2 O3
Certifikat och efterlevnad ISO 17025
Synonym Flutamide Imp. C (EP), N-[4-Nitro-3-(trifluoromethyl)phenyl]propanamide, Flutamide Impurity C
InChI formel InChI=1S/C10H9F3N2O3/c1-2-9(16)14-6-3-4-8(15(17)18)7(5-6)10(11,12)13/h3-5H,2H2,1H3,(H,14,16)
LEDER CCC(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC-namn N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Molekylvikt (g/mol) 262.19
Kvantitet 25 mg
Formel vikt 262.0565 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.