Läs mer
N-(4-Nitrobenzoyl)-L-glutamic Acid, TRC
Produktkod. 30475745
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30475745 25 g
1 options
Produktkod. 30475745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49368525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(4-Nitrobenzoyl)-L-glutamic Acid
CAS 6758-40-3
Molekylformel C12 H12 N2 O7
Kvantitet 25 g
Synonym N-(p-Nitrobenzoyl)-L(+)-glutamic Acid, NSC 17156, NSC 83096, p-Nitrobenzoyl-L-glutamic Acid, p-Nitrobenzoylglutamic Acid
InChI formel InChI=1S/C12H12N2O7/c15-10(16)6-5-9(12(18)19)13-11(17)7-1-3-8(4-2-7)14(20)21/h1-4,9H,5-6H2,(H,13,17)(H,15,16)(H,18,19)/t9-/m0/s1
LEDER OC(=O)CC[C@H](NC(=O)c1ccc(cc1)[N+](=O)[O-])C(=O)O
IUPAC-namn (2S)-2-[(4-nitrobenzoyl)amino]pentanedioic acid
Molekylvikt (g/mol) 296.23
Formel vikt 296.0645