Läs mer
N-(5-Bromo-6-quinoxalinyl)quanidine Hydrochloride, TRC
Produktkod. 30180391
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180391 100 mg
30179898 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180391 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB687050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797986-78-7
Molekylformel C9 H8 Br N5 . Cl H
Molekylvikt (g/mol) 302.56
Synonym Guanidine, N-(5-bromo-6-quinoxalinyl)-, hydrochloride (1:1), N-(5-Bromo-6-quinoxalinyl)guanidine hydrochloride, 2-(5-Bromoquinoxalin-6-yl)guanidine hydrochloride
IUPAC-namn 2-(5-bromoquinoxalin-6-yl)guanidine;hydrochloride
LEDER Cl.NC(=Nc1ccc2nccnc2c1Br)N
Specifikationer
Kemiskt namn eller material 2-(5-Bromoquinoxalin-6-yl)guanidine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C9H8BrN5.ClH/c10-7-5(15-9(11)12)1-2-6-8(7)14-4-3-13-6;/h1-4H,(H4,11,12,15);1H
Formel vikt 300.973
Procent renhet >95%