Learn More
Alfa Aesar™ N-(5-Chloro-2-methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%
Brand:  Alfa Aesar™ H65868.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15499956
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 05-03-2018
Add to basket
Specifications
Chemical Identifiers
N-(5-Chloro-2-methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
299.8
MFCD27975834
Specifications
N-(5-Chloro-2-methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
95%
MFCD27975834
Solid
299.8
1g