Läs mer
N-[5-(Hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl]-carbamic Acid Benzyl Ester, TRC
Produktkod. 30404055
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30404055 100 mg
1 options
Produktkod. 30404055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH948015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[5-(Hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl]-carbamic Acid Benzyl Ester
CAS 1125824-87-4
Molekylformel C19H21NO5
Kvantitet 100 mg
Synonym N-[5-(Hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl]-carbamic Acid Phenylmethyl Ester
InChI formel InChI=1S/C19H21NO5/c21-12-19(20-18(22)23-11-15-7-3-1-4-8-15)13-24-17(25-14-19)16-9-5-2-6-10-16/h1-10,17,21H,11-14H2,(H,20,22)
LEDER O=C(OCC1=CC=CC=C1)NC2(CO)COC(C3=CC=CC=C3)OC2
IUPAC-namn benzyl (5-(hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl)carbamate
Molekylvikt (g/mol) 343.37
Formel vikt 243.14